The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293622.1 | NP_001280551.1 | 1 - 2 | 838 | 72 |
NM_008913.5 | NP_032939.1 | 1 - 2 | 838 | 72 | |
GenBank mRNA | AK146387.1 | - | 1 - 2 | 782 | 72 |
AK150393.1 | - | 1 - 2 | 197 | 72 | |
AK166287.1 | - | 1 - 2 | 613 | 72 | |
BC096652.1 | - | 1 - 2 | 80 | 72 | |
BC138612.1 | - | 1 - 2 | 111 | 72 | |
J05479.1 | - | 1 - 2 | 135 | 72 |