The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053080.3 | NP_444310.3 | 11 - 12 | 1459 | 93 |
GenBank mRNA | AF152359.1 | - | 11 - 12 | 1414 | 93 |
AF246711.1 | - | 11 - 12 | 1407 | 93 | |
AF253409.1 | - | 11 - 12 | 1446 | 93 | |
AF280404.1 | - | 11 - 12 | 1411 | 93 | |
AK147006.1 | - | 11 - 12 | 1460 | 93 | |
BC038646.1 | - | 2 - 3 | 280 | 93 | |
BC048941.1 | - | 3 - 4 | 372 | 93 | |
BC058277.1 | - | 11 - 12 | 1403 | 93 |