Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo16
Entrez Gene ID
50759
Gene Name
F-box protein 16
Gene Aliases
4932435C24Rik, Fbx16
Chromosome Location
Chr.14: 65266123 - 65323655 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015795.1 NP_056610.1 6 - 7 856 107
GenBank mRNA AF176531.1 - 6 - 7 856 107
AK043554.1 - 6 - 7 826 107
AK164657.1 - 6 - 7 939 107
BC086690.1 - 6 - 7 820 107
BC115890.1 - 6 - 7 814 107

View Full Product Details