Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grm5
Entrez Gene ID
108071
Gene Name
glutamate receptor, metabotropic 5
Gene Aliases
6430542K11Rik, AI850523, Glu5R, Gprc1e, mGluR5, mGluR5b
Chromosome Location
Chr.7: 87583981 - 88135063 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143834.1 NP_001137306.1 9 - 10 3343 73
GenBank mRNA BC096533.1 - 7 - 8 2728 73
BC144727.1 - 6 - 7 1968 73
EF508472.1 - 7 - 8 2625 73

View Full Product Details