Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap1ar
Entrez Gene ID
211556
Gene Name
adaptor-related protein complex 1 associated regulatory protein
Gene Aliases
AA407621, C78909, Gadkin
Chromosome Location
Chr.3: 127807007 - 127837518 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145964.2 NP_666076.2 9 - 10 894 64
GenBank mRNA BB611022.1 - 9 - 10 582 64

View Full Product Details