The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243047.1 | NP_001229976.1 | 6 - 7 | 713 | 100 |
NM_011807.3 | NP_035937.2 | 18 - 19 | 2189 | 100 | |
GenBank mRNA | AF388675.1 | - | 18 - 19 | 1968 | 100 |
AK039754.1 | - | 6 - 7 | 578 | 100 |