Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kif7
Entrez Gene ID
16576
Gene Name
kinesin family member 7
Gene Aliases
-
Chromosome Location
Chr.7: 79698098 - 79715772 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291222.1 NP_001278151.1 15 - 16 3358 70
NM_010626.3 NP_034756.2 15 - 16 3355 70
GenBank mRNA BC034093.1 - 5 - 6 930 70
BC141045.1 - 15 - 16 3356 70
BC145222.1 - 15 - 16 3359 70

View Full Product Details