Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Synm
- Entrez Gene ID
-
233335
- Gene Name
- synemin, intermediate filament protein
- Gene Aliases
-
4930412K21Rik, AI852401, Dmn, E130104F11, Syn, Synemin
- Chromosome Location
-
Chr.7: 67730161 - 67759742 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK053514.1 |
- |
1 - 2 |
70 |
102 |
View Full Product Details