The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033877.4 | NP_001029049.2 | 6 - 7 | 1132 | 67 |
GenBank mRNA | AK045148.1 | - | 7 - 8 | 1253 | 67 |
AK053131.1 | - | 6 - 7 | 1133 | 67 | |
BC141170.1 | - | 6 - 7 | 1084 | 67 |