Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
l7Rn6
Entrez Gene ID
67669
Gene Name
lethal, Chr 7, Rinchik 6
Gene Aliases
0610007P06Rik, 1110002N09Rik, Hikeshi, l(7)6Rn
Chromosome Location
Chr.7: 89918685 - 89941221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291288.1 NP_001278217.1 3 - 4 616 74
GenBank mRNA BC083330.1 - 3 - 4 477 74

View Full Product Details