The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080443.2 | NP_536691.2 | 5 - 6 | 1532 | 60 |
NR_003961.1 | 7 - 8 | 1696 | 60 | ||
GenBank mRNA | BC062948.1 | - | 3 - 4 | 850 | 60 |
BC066824.1 | - | 5 - 6 | 1123 | 60 | |
BC085155.1 | - | 5 - 6 | 1567 | 60 |