The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080728.2 | NP_542766.1 | 35 - 36 | 5095 | 66 |
GenBank mRNA | AK147031.1 | - | 35 - 36 | 5096 | 66 |
AK168593.1 | - | 16 - 17 | 2864 | 66 | |
AK168677.1 | - | 19 - 20 | 3126 | 66 | |
AK169198.1 | - | 35 - 36 | 5095 | 66 | |
AK169371.1 | - | 35 - 36 | 5095 | 66 | |
AY056464.1 | - | 35 - 36 | 5095 | 66 | |
BC121789.1 | - | 34 - 35 | 5051 | 66 | |
BC158018.1 | - | - | 5172 | 66 | |
BC158070.1 | - | 35 - 36 | 5350 | 66 | |
M74752.1 | - | 10 - 11 | 1748 | 66 |