Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Wdfy2
- Entrez Gene ID
-
268752
- Gene Name
- WD repeat and FYVE domain containing 2
- Gene Aliases
-
5830485M08, B130024L21Rik, ProF, ZFYVE22
- Chromosome Location
-
Chr.14: 62837690 - 62961509 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK045072.1 |
- |
2 - 3 |
240 |
105 |
View Full Product Details