Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cysltr2
Entrez Gene ID
70086
Gene Name
cysteinyl leukotriene receptor 2
Gene Aliases
2300001H05Rik, CYSLT2R, Cltr2, Cyslt2
Chromosome Location
Chr.14: 73026075 - 73049114 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162412.1 NP_001155884.1 2 - 3 185 155
GenBank mRNA AK008997.1 - 2 - 3 186 155

View Full Product Details