The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163422.1 | NP_001156894.1 | 8 - 9 | 635 | 62 |
NM_026895.1 | NP_081171.1 | 9 - 10 | 748 | 62 | |
GenBank mRNA | AK005193.1 | - | 8 - 9 | 606 | 62 |
AK156071.1 | - | 9 - 10 | 699 | 62 | |
BC094396.1 | - | 4 - 5 | 278 | 62 |