Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnih3
Entrez Gene ID
72978
Gene Name
cornichon homolog 3 (Drosophila)
Gene Aliases
2900075G08Rik, CNIH-3
Chromosome Location
Chr.1: 181352628 - 181460641 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160211.1 NP_001153683.1 3 - 4 996 65
NM_001160212.1 NP_001153684.1 3 - 4 996 65
GenBank mRNA AK033911.1 - 3 - 4 994 65
BC115640.1 - 3 - 4 622 65
BC115641.1 - 3 - 4 622 65

View Full Product Details