Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem55b
Entrez Gene ID
219024
Gene Name
transmembrane protein 55b
Gene Aliases
BC039161
Chromosome Location
Chr.14: 50926070 - 50930905 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033271.4 NP_001028443.1 3 - 4 536 68
GenBank mRNA AK144251.1 - 3 - 4 536 68
AK152640.1 - 3 - 4 522 68
AK152658.1 - 3 - 4 522 68
AK159641.1 - 3 - 4 540 68
BC027355.1 - 3 - 4 521 68
BC058171.1 - 3 - 4 485 68
BC106163.1 - 3 - 4 515 68

View Full Product Details