Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rarb
Entrez Gene ID
218772
Gene Name
retinoic acid receptor, beta
Gene Aliases
A830025K23, Hap, Nr1b2
Chromosome Location
Chr.14: 16430839 - 17082331 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011243.2 NP_035373.1 1 - 2 621 64
GenBank mRNA AK043726.1 - 1 - 2 974 64
AK054178.1 - 1 - 2 585 64
AK078471.1 - 1 - 2 334 64
BC076597.1 - 1 - 2 617 64
S56660.1 - 1 - 2 517 64

View Full Product Details