Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gbp5
Entrez Gene ID
229898
Gene Name
guanylate binding protein 5
Gene Aliases
5330409J06Rik, Gbp5a
Chromosome Location
Chr.3: 142496934 - 142522344 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF487898.1 - 9 - 10 2315 63
AK030414.1 - 11 - 12 2659 63

View Full Product Details