Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc28
Entrez Gene ID
67867
Gene Name
leucine rich repeat containing 28
Gene Aliases
1300004K21Rik, 2210012C09Rik, 2310058O11Rik
Chromosome Location
Chr.7: 67513410 - 67645331 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028144.1 3 - 4 340 101
GenBank mRNA BC052051.1 - 2 - 3 247 101

View Full Product Details