Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oxa1l
Entrez Gene ID
69089
Gene Name
oxidase assembly 1-like
Gene Aliases
1810020M02Rik, AI894287
Chromosome Location
Chr.14: 54360841 - 54369669 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026936.3 NP_081212.1 5 - 6 668 126
GenBank mRNA AK028379.1 - 5 - 6 667 126
AK044817.1 - 5 - 6 667 126
AK052432.1 - 5 - 6 669 126
AK077624.1 - 5 - 6 669 126
AK087756.1 - 5 - 6 672 126
AK139484.1 - 5 - 6 676 126
AK154304.1 - 5 - 6 671 126
AK154532.1 - 5 - 6 669 126
AK170898.1 - 5 - 6 669 126
BC027191.1 - 6 - 7 600 126

View Full Product Details