Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Synpr
Entrez Gene ID
72003
Gene Name
synaptoporin
Gene Aliases
1500003F20Rik, SPO
Chromosome Location
Chr.14: 13284780 - 13615469 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028052.4 NP_082328.3 2 - 3 290 88
GenBank mRNA AK005132.1 - 2 - 3 211 88
AK078223.1 - 2 - 3 291 88
BC026512.1 - 2 - 3 237 88

View Full Product Details