Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smyd3
Entrez Gene ID
69726
Gene Name
SET and MYND domain containing 3
Gene Aliases
2410008A19Rik, Zmynd1
Chromosome Location
Chr.1: 178951960 - 179518052 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027188.3 NP_081464.1 1 - 2 214 63
GenBank mRNA AK010447.1 - 1 - 2 248 63
AK043988.1 - 1 - 2 225 63
AK087728.1 - 1 - 2 225 63
AK163057.1 - 1 - 2 231 63
BC019663.1 - 1 - 2 216 63
BC038482.1 - 1 - 2 95 63
BC052431.1 - 1 - 2 214 63
BC061485.1 - 1 - 2 197 63

View Full Product Details