Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adh4
Entrez Gene ID
26876
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Gene Aliases
Adh2
Chromosome Location
Chr.3: 138415497 - 138430892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011996.2 NP_036126.2 6 - 7 874 93
GenBank mRNA AJ245750.1 - 6 - 7 862 93
AK132994.1 - 6 - 7 874 93
BC100729.1 - 6 - 7 862 93
BC100730.1 - 6 - 7 861 93
BC100736.1 - 6 - 7 862 93

View Full Product Details