Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpe65
Entrez Gene ID
19892
Gene Name
retinal pigment epithelium 65
Gene Aliases
65kDa, A930029L06Rik, LCA2, Mord1, RP20, rd12
Chromosome Location
Chr.3: 159599075 - 159625307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029987.2 NP_084263.2 9 - 10 1045 169
GenBank mRNA AF410461.1 - 8 - 9 993 169
BC090656.1 - 9 - 10 1021 169
BC130028.1 - 8 - 9 997 169

View Full Product Details