Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss12
Entrez Gene ID
19142
Gene Name
protease, serine 12 neurotrypsin (motopsin)
Gene Aliases
Bssp-3, motopsin
Chromosome Location
Chr.3: 123446913 - 123506602 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008939.2 NP_032965.1 4 - 5 1201 86
GenBank mRNA AK132633.1 - 4 - 5 990 86
BC031429.1 - 4 - 5 1103 86
D89871.1 - 4 - 5 1199 86
Y13192.1 - 4 - 5 978 86

View Full Product Details