The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291222.1 | NP_001278151.1 | 16 - 17 | 3561 | 81 |
NM_010626.3 | NP_034756.2 | 16 - 17 | 3558 | 81 | |
GenBank mRNA | BC034093.1 | - | 6 - 7 | 1133 | 81 |
BC141045.1 | - | 16 - 17 | 3559 | 81 | |
BC145222.1 | - | 16 - 17 | 3562 | 81 |