Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plin1
Entrez Gene ID
103968
Gene Name
perilipin 1
Gene Aliases
6030432J05Rik, Peri, Plin
Chromosome Location
Chr.7: 79721164 - 79732904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113471.1 NP_001106942.1 7 - 8 1020 150
NM_175640.2 NP_783571.2 7 - 8 1076 150
GenBank mRNA AK031445.1 - 6 - 7 1001 150
AY161165.1 - 6 - 7 971 150
BC096685.1 - 7 - 8 1021 150

View Full Product Details