Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smyd2
Entrez Gene ID
226830
Gene Name
SET and MYND domain containing 2
Gene Aliases
1110020E07Rik, 4930402C15, KMT3C, Zmynd14
Chromosome Location
Chr.1: 189880492 - 189922379 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026796.1 NP_081072.1 1 - 2 187 118
GenBank mRNA AK145261.1 - 1 - 2 192 119
AK150857.1 - 1 - 2 191 118
AK164053.1 - 1 - 2 153 118
AK204951.1 - 1 - 2 95 118
BC023119.1 - 1 - 2 187 118

View Full Product Details