The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026171.1 | NP_080447.1 | 13 - 14 | 1627 | 55 |
GenBank mRNA | AK004676.1 | - | 13 - 14 | 1659 | 55 |
AK054502.1 | - | 8 - 9 | 1106 | 55 | |
AK140509.1 | - | 13 - 14 | 1658 | 55 | |
BC031847.1 | - | 13 - 14 | 1610 | 55 |