The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015771.2 | NP_056586.2 | 1 - 2 | 231 | 137 |
GenBank mRNA | AK029242.1 | - | - | 212 | 137 |
AK029870.1 | - | 1 - 2 | 212 | 137 | |
AK029966.1 | - | 1 - 2 | 191 | 137 | |
AK081190.2 | - | 1 - 2 | 230 | 137 | |
AK163607.1 | - | 1 - 2 | 214 | 137 | |
BC024819.2 | - | 1 - 2 | 222 | 137 | |
BC053028.1 | - | 1 - 2 | 185 | 137 | |
BU705730.1 | - | 1 - 2 | 231 | 137 |