The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020490.2 | NP_065236.1 | 2 - 2 | 1268 | 63 |
GenBank mRNA | AK018029.1 | - | 2 - 2 | 1260 | 63 |
AK036836.1 | - | - | 1628 | 63 |