The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027514.2 | NP_081790.1 | 1 - 2 | 195 | 59 |
GenBank mRNA | AB195430.1 | - | 1 - 2 | 152 | 59 |
AJ563420.1 | - | 1 - 2 | 83 | 59 | |
AK014446.1 | - | 1 - 2 | 166 | 59 | |
AK038747.1 | - | 1 - 2 | 166 | 59 | |
AK040334.1 | - | 1 - 2 | 166 | 59 | |
AK078344.1 | - | 1 - 2 | 162 | 59 | |
AK080753.1 | - | 1 - 2 | 166 | 59 | |
AK165873.1 | - | 1 - 2 | 185 | 59 | |
BC013673.1 | - | 1 - 2 | 152 | 59 | |
BC032283.1 | - | 1 - 2 | 155 | 59 | |
BC138219.1 | - | 1 - 2 | 140 | 59 | |
BC138220.1 | - | 1 - 2 | 140 | 59 | |
BY276828.1 | - | 1 - 2 | 196 | 59 |