Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcl3
Entrez Gene ID
12051
Gene Name
B cell leukemia/lymphoma 3
Gene Aliases
AI528691, Bcl-3
Chromosome Location
Chr.7: 19808462 - 19822811 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033601.3 NP_291079.2 2 - 3 476 135
GenBank mRNA AF067774.1 - 2 - 3 437 135
AK155016.1 - 2 - 3 441 135
BY749796.1 - 2 - 3 477 135
M90397.1 - 2 - 3 394 135

View Full Product Details