Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc44a3
Entrez Gene ID
213603
Gene Name
solute carrier family 44, member 3
Gene Aliases
BC010552
Chromosome Location
Chr.3: 121459528 - 121532344 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145394.3 NP_663369.2 2 - 3 293 70
GenBank mRNA BC010552.1 - 1 - 2 61 70
BC025548.1 - 2 - 3 291 70
CJ094004.1 - 2 - 3 236 70

View Full Product Details