Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmp4
Entrez Gene ID
12159
Gene Name
bone morphogenetic protein 4
Gene Aliases
Bmp-4, Bmp2b, Bmp2b-1, Bmp2b1
Chromosome Location
Chr.14: 46383520 - 46390599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007554.2 NP_031580.2 1 - 2 291 57
GenBank mRNA AK136527.1 - 1 - 2 292 57
AK145359.1 - 1 - 2 361 57
BC013459.1 - 1 - 2 263 57
X56848.1 - 1 - 2 219 57

View Full Product Details