The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289729.1 | NP_001276658.1 | 5 - 6 | 828 | 78 |
NM_133213.2 | NP_573476.2 | 5 - 6 | 828 | 78 | |
GenBank mRNA | AF367247.1 | - | 5 - 6 | 492 | 78 |
AF428102.1 | - | 5 - 6 | 679 | 78 | |
AK078835.1 | - | 5 - 6 | 635 | 78 | |
AK085520.1 | - | 5 - 6 | 626 | 78 | |
AK087627.1 | - | 5 - 6 | 635 | 78 | |
AK164538.1 | - | 5 - 6 | 656 | 78 | |
AK172086.1 | - | 5 - 6 | 829 | 78 | |
BC140977.1 | - | 5 - 6 | 472 | 78 |