The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290364.1 | NP_001277293.1 | 14 - 15 | 1777 | 65 |
NM_012019.3 | NP_036149.1 | 14 - 15 | 1789 | 65 | |
GenBank mRNA | AF100927.1 | - | 14 - 15 | 1615 | 65 |
AK133764.1 | - | 14 - 15 | 1629 | 65 | |
AK165762.1 | - | 4 - 5 | 580 | 65 | |
AK219625.1 | - | 2 - 3 | 152 | 65 | |
BC003292.1 | - | 14 - 15 | 1652 | 65 | |
BC024882.1 | - | 14 - 15 | 2273 | 65 | |
DQ016499.1 | - | 13 - 14 | 2916 | 65 |