Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankib1
Entrez Gene ID
70797
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
2310061P20Rik, 4631416I11Rik, AW494740, C80642, mKIAA1386
Chromosome Location
Chr.5: 3689999 - 3803124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003909.4 NP_001003909.2 13 - 14 2456 113
NM_001289527.1 NP_001276456.1 13 - 14 2456 113
NM_001289528.1 NP_001276457.1 12 - 13 2338 113
NM_001289529.1 NP_001276458.1 13 - 14 2065 113
GenBank mRNA AK129343.1 - 13 - 14 2257 113
BC020475.1 - 13 - 14 1990 113
BC089031.1 - 13 - 14 2044 113
BC145510.1 - 12 - 13 2089 113
BC145511.1 - 13 - 14 2207 113
BC150792.1 - 13 - 14 2207 113

View Full Product Details