Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdc42bpa
Entrez Gene ID
226751
Gene Name
CDC42 binding protein kinase alpha
Gene Aliases
A930014J19Rik, DMPK-like
Chromosome Location
Chr.1: 179960093 - 180165606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033285.1 NP_001028457.1 5 - 6 627 110
GenBank mRNA AK138649.1 - 6 - 7 1116 110
BC158017.1 - 5 - 6 627 110
BC158095.1 - 5 - 6 627 110

View Full Product Details