Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gmfb
- Entrez Gene ID
-
63985
- Gene Name
- glia maturation factor, beta
- Gene Aliases
-
3110001H22Rik, 3110001O16Rik, AI851627, C79176, D14Ertd630e
- Chromosome Location
-
Chr.14: 46808149 - 46822263 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB001732.1 |
- |
3 - 4 |
222 |
67 |
View Full Product Details