The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011892.3 | NP_036022.1 | 2 - 3 | 343 | 86 |
GenBank mRNA | AF282901.1 | - | 2 - 3 | 343 | 86 |
AK132610.1 | - | 2 - 3 | 288 | 86 | |
BC138960.1 | - | 2 - 3 | 235 | 86 | |
BC138961.1 | - | 2 - 3 | 235 | 86 |