Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf6
Entrez Gene ID
70998
Gene Name
PHD finger protein 6
Gene Aliases
2700007B13Rik, 4931428F02Rik, mKIAA1823
Chromosome Location
Chr.X: 52912214 - 52956961 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027642.2 NP_081918.1 3 - 4 511 123
GenBank mRNA AK016480.1 - 3 - 4 512 123
AK038320.1 - 3 - 4 431 123
AK122560.1 - 3 - 4 441 123
AK141804.1 - 3 - 4 454 123
AK163345.1 - 3 - 4 467 123
AK213676.1 - 3 - 4 349 123
BC043127.1 - 3 - 4 459 123
BC055330.1 - 3 - 4 454 123
BC057374.1 - 3 - 4 417 123

View Full Product Details