Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phf6
Entrez Gene ID
70998
Gene Name
PHD finger protein 6
Gene Aliases
2700007B13Rik, 4931428F02Rik, mKIAA1823
Chromosome Location
Chr.X: 52912214 - 52956961 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290546.1 NP_001277475.1 2 - 3 354 78
NM_027642.2 NP_081918.1 4 - 5 640 78
GenBank mRNA AK016480.1 - 4 - 5 641 78
AK028498.1 - 2 - 3 276 78
AK038320.1 - 4 - 5 560 78
AK122560.1 - 4 - 5 570 78
AK141804.1 - 4 - 5 583 78
AK163345.1 - 4 - 5 596 78
BC043127.1 - 4 - 5 588 78
BC055330.1 - 4 - 5 583 78
BC057374.1 - 4 - 5 546 78

View Full Product Details