Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcpt2
Entrez Gene ID
17225
Gene Name
mast cell protease 2
Gene Aliases
MMCP-2, Mcp-2
Chromosome Location
Chr.14: 56021584 - 56044635 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008571.1 NP_032597.1 2 - 3 243 107
GenBank mRNA AK133521.1 - 2 - 3 244 107
AY007570.1 - 2 - 3 243 107
BC119248.1 - 2 - 3 236 107
BC119250.1 - 2 - 3 236 107
J05177.1 - 2 - 3 107

View Full Product Details