Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Igsf1
Entrez Gene ID
209268
Gene Name
immunoglobulin superfamily, member 1
Gene Aliases
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364
Chromosome Location
Chr.X: 49782536 - 49797906 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177591.4 NP_808259.2 6 - 7 1101 73
NM_183335.2 NP_899178.2 5 - 6 965 73
GenBank mRNA AK129126.1 - 6 - 7 1083 73
AY227771.1 - 6 - 7 1095 73
AY227773.1 - 5 - 6 965 73

View Full Product Details