Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a40
Entrez Gene ID
319653
Gene Name
solute carrier family 25, member 40
Gene Aliases
B230315F11Rik
Chromosome Location
Chr.5: 8422838 - 8454840 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK163037.1 - 1 - 2 172 78

View Full Product Details