Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc61
Entrez Gene ID
232933
Gene Name
coiled-coil domain containing 61
Gene Aliases
C530028I08Rik, Gm159
Chromosome Location
Chr.7: 18890883 - 18910412 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033314.3 NP_001028486.1 2 - 3 423 82
GenBank mRNA AK082980.1 - 2 - 3 418 82
AK146297.1 - 2 - 3 424 82

View Full Product Details