Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fibcd1
Entrez Gene ID
98970
Gene Name
fibrinogen C domain containing 1
Gene Aliases
AI448887
Chromosome Location
Chr.2: 31813290 - 31846647 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178887.4 NP_849218.2 1 - 2 291 73
GenBank mRNA AK084379.1 - 1 - 2 256 73
BC060634.1 - 1 - 2 291 73

View Full Product Details