Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abl1
Entrez Gene ID
11350
Gene Name
c-abl oncogene 1, non-receptor tyrosine kinase
Gene Aliases
AI325092, Abl, E430008G22Rik, c-Abl
Chromosome Location
Chr.2: 31688252 - 31807093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112703.2 NP_001106174.1 3 - 4 1879 112
NM_001283045.1 NP_001269974.1 3 - 4 756 112
NM_001283046.1 NP_001269975.1 4 - 5 2084 112
NM_001283047.1 NP_001269976.1 4 - 5 2023 112
NM_009594.4 NP_033724.2 3 - 4 650 112
GenBank mRNA AK088252.1 - 4 - 5 2009 112
AK090095.1 - 3 - 4 608 112
AK166028.1 - 3 - 4 1868 112
AK166210.1 - 4 - 5 2118 112
BC059260.1 - 3 - 4 677 112
BC103770.1 - 3 - 4 1103 112
CF536749.1 - 1 - 2 254 112
CN663033.1 - 2 - 3 371 112
J02995.1 - 3 - 4 603 112
L10656.1 - 1 - 2 204 112

View Full Product Details